2-Amino-3-hydroxypyridin 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Aminopyridin-3-ol, 2-Amino-3-hydroxypyridine

A10665.18 A10665.09 A10665.22 A10665.30
A10665.18EA 882 DKK
A10665.18 A10665.09 A10665.22 A10665.30
2-Amino-3-hydroxypyridin 98%
2-Amino-3-hydroxypyridin

Purity: 98% CAS No.: 16867-03-1 Molecular Formula: C5H6N2O Formula Weight: 110.12 Odour: Odourless Melting Point: 166-174° Beilstein Reference: 109868 UN No.: 2811 MDL No.: MFCD00006317

Formel: C₅H₆N₂O
Kogepunkt: 250 °C (1013 hPa)
Smeltepunkt: 166…174 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006317
CAS-nummer: 16867-03-1
EINECS: 240-886-8
UN nummer: 2811
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR