Pyridin-2-caboxylsyre 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Pyridine-2-carboxylic acid, 2-Picolinic acid, 2-Pyridinecarboxylic acid
131170250
131171000
131175000
131170025
ACRO131170250EA
209
DKK
ACRO131170250
ACRO131171000
ACRO131175000
ACRO131170025
Pyridin-2-caboxylsyre 99%
Pyridin-2-caboxylsyre
Formel:
C₆H₅NO₂ Kogepunkt: 293 °C (1013 hPa) Smeltepunkt: 136…139 °C Massefylde: 1,49 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006293 CAS-nummer: 98-98-6 EINECS: 202-719-7 Merck Index: 12,07557 |
Specifikationer Testresultater
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 136 to 139 °C |
Titration with NaOH | 98.5 to 101.5 % |
Residue after evaporation | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...