Pyridin-2-carbonitril 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Cyanopyridine, 2-Pyridinecarbonitrile, Picolinonitrile

110850050 110851000 110855000
ACRO110850050EA 276 DKK
ACRO110850050 ACRO110851000 ACRO110855000
Pyridin-2-carbonitril 99%
Pyridin-2-carbonitril
Formel: C₆H₄N₂
Kogepunkt: 212 °C (1013 hPa)
Smeltepunkt: 26…28 °C
Massefylde: 1,081 g/cm³ (25 °C)
Flammepunkt: 89 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006218
CAS-nummer: 100-70-9
EINECS: 202-880-3
UN nummer: 3439

Bestil nu

Specifikationer Testresultater

Appearance White to brown Fused solid or liquid
Infrared spectrum Conforms
Melting point 25 to 31 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR