2-Chlor-3-fluorpyridin ≥97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
L20022.03
L20022.06
L20022.03EA
406
DKK
L20022.03
L20022.06
2-Chlor-3-fluorpyridin ≥97%
2-Chloro-3-fluoropyridine
Purity: 97%
CAS No.: 17282-04-1
Molecular Formula: C5H3ClFN
Formula Weight: 131.54
Flash Point: 72°(161°F)
Refractive Index: 1.5095
Odour: Unpleasant
Boiling Point: 62-64°/23mm
Beilstein Reference: 1363435
UN No.: 2810
MDL No.: MFCD03095302
Formel:
C₅H₃ClFN Kogepunkt: 62…64 °C (23 mmHg) Massefylde: 1,325 Flammepunkt: 72 °C (161 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD03095302 CAS-nummer: 17282-04-1 UN nummer: 2810 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...