5-Brom-2-fluorpyridin ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

L19537.06 L19537.14
L19537.06EA 1150 DKK
L19537.06 L19537.14
5-Brom-2-fluorpyridin ≥98%
5-Brom-2-fluorpyridin

Purity: 98% CAS No.: 766-11-0 Molecular Formula: C5H3BrFN Formula Weight: 175.99 Flash Point: 73°(163°F) Refractive Index: 1.5294 Odour: Characteristic Boiling Point: 162-164° Beilstein Reference: 1363171 UN No.: 2810 MDL No.: MFCD01863742

Formel: C₅H₃BrFN
Molekylvægt: 175.99 g/mol
Kogepunkt: 162…164 °C
Massefylde: 1.72
Flammepunkt: 73 °C (163 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01863742
CAS-nummer: 766-11-0
UN nummer: 2810
ADR: 6.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR