1-Aminopyren 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

104530010 104530050
ACRO104530010EA 696 DKK
ACRO104530010 ACRO104530050
1-Aminopyren 97%
1-Aminopyrene
Formel: C₁₆H₁₁N
Kogepunkt: 415 °C (1013 hPa)
Smeltepunkt: 114…118 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004140
CAS-nummer: 1606-67-3
EINECS: 216-521-3

Bestil nu

Specifikationer Testresultater

Appearance Yellow to green Powder
Infrared spectrum Conforms
Melting point 114 to 118 °C
Titration with HClO4 ≥96.0 %
Average molecular weight 70000 (Mn)
Acetyl content 12 to 15 %
Hydroxyl content 1.8 % Typical
Butyryl content 35 to 39 %
Free acid (CH3COOH) ≤0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR