Coffein, EMPROVE® ESSENTIAL Ph. Eur., USP, BP suitable for use as excipient, SAFC®
Leverandør: MERCK PRODUCTION CHEMICALS
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 7-Methyltheobromine, 1,3,7-Trimethylxanthine, Methyltheobromine, 1,3,7-Trimethyl-2,6-dioxopurine, 2,6-Dihydroxy-1,3,7-trimethylpurine
1.02584.1000
1.02584.9050
1.02584.1000EA
5060
DKK
1.02584.1000
1.02584.9050
Coffein, EMPROVE® ESSENTIAL Ph. Eur., USP, BP suitable for use as excipient, SAFC®
Coffein
Formel:
C₈H₁₀N₄O₂ Molekylvægt: 194.19 g/mol Smeltepunkt: 235…239 °C Massefylde: 1.23 g/cm³ (19 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00005758 CAS-nummer: 58-08-2 EINECS: 200-362-1 Merck Index: 13,01636 |