Nikkelmonoxid 97%, powder

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Bunsenit, Nikkeloxid

415585000 415581000
ACRO415585000EA 1510 DKK
ACRO415585000 ACRO415581000
Nikkelmonoxid 97%, powder
Nikkelmonoxid
Formel: NiO
Smeltepunkt: 1960 °C
Massefylde: 4,6 to 6.67 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011145
CAS-nummer: 1313-99-1
EINECS: 215-215-7

Bestil nu

Specifikationer Testresultater

Appearance Green Powder
Titration Complexometric ≥96.0 %
Calcium (Ca) ≤0.003 %
Cobalt (Co) ≤0.025 %
Copper (Cu) ≤0.0005 %
Iron (Fe) ≤0.005 %
Magnesium (Mg) ≤0.01 %
Manganese (Mn) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR