Mangan(IV)oxid ≥98%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Mangandioxid
A10765.36 A10765.A9 A10765.0E A10765.09
A10765.36EA 996 DKK
A10765.36 A10765.A9 A10765.0E A10765.09
Mangan(IV)oxid ≥98%
Manganese(IV) oxide
Purity: 98% CAS No.: 1313-13-9 Molecular Formula: MnO2 Formula Weight: 86.94 Melting Point: 535° dec. Merck Reference: 14,5730 MDL No.: MFCD00003463
Formel: MnO₂
Molekylvægt: 86.94 g/mol
Smeltepunkt: 535 °C (dec.)
Massefylde: 5.026
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003463
CAS-nummer: 1313-13-9
EINECS: 215-202-6
Merck Index: 14,5730