Mangan(IV)oxid 99.99+% (trace metals basis)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Mangandioxid
193470050 193470250 193471000
ACRO193470050EA 557 DKK
ACRO193470050 ACRO193470250 ACRO193471000
Mangan(IV)oxid 99.99+% (trace metals basis)
Manganese(IV) oxide
Formel: MnO₂
Molekylvægt: 86.94 g/mol
Smeltepunkt: 535 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003463
CAS-nummer: 1313-13-9
EINECS: 215-202-6
Merck Index: 14,05730
Bestil nu
Specifikationer Testresultater
Appearance Grey to black Powder or crystals and/or chunks
Total trace metal impurities ≤100 ppm