Bottles, wide neck, LDPE
Leverandør: RIXIUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
U3S0310005032 U3S0310010032 U3S0310025040 U3S0310050050 U3S0310100065 U3S0310200065
RIXIU3S0310005032EA 8110 DKK
RIXIU3S0310005032 RIXIU3S0310010032 RIXIU3S0310025040 RIXIU3S0310050050 RIXIU3S0310100065 RIXIU3S0310200065
Bottles, wide neck, LDPE
Flasker Plastflasker
LDPE, natural, with screw caps.
  • Resistant to acids, alkalis and other chemicals
  • Temperature resistant: –20 to +70 °C
Perfect for sample collection, storage and transport. Ideal for paste-like media and granular material. The wide mouth allows easy filling and cleaning.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904