Cadmium (II) carbonat, pure

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

212320010 212322500
ACRO212320010EA 2040 DKK
ACRO212320010 ACRO212322500
Cadmium (II) carbonat, pure
Cadmium (II) carbonat
Formel: CCdO₃
Molekylvægt: 172.42 g/mol
Smeltepunkt: 343…412 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010918
CAS-nummer: 513-78-0
EINECS: 208-168-9
UN nummer: 2570
ADR: 6.1 II
Merck Index: 12,01652

Bestil nu

Specifikationer Testresultater

Appearance White to off-white Powder
Assay 60.0 to 63.0 % (Cd)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR