Kobber(II)sulfat, anhydrous 98%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Cupric sulphate
A13986.0B A13986.0I A13986.30
A13986.0BEA 1740 DKK
A13986.0B A13986.0I A13986.30
Kobber(II)sulfat, anhydrous 98%
Kobber(II)sulfat
Purity: 98% CAS No.: 7758-98-7 Molecular Formula: CuO4S Formula Weight: 159.60 Odour: Odourless Melting Point: 560° UN No.: 3288 MDL No.: MFCD00010981
Formel: CuSO₄
Molekylvægt: 159.61 g/mol
Smeltepunkt: >560 °C
Massefylde: 3.6
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010981
CAS-nummer: 7758-98-7
EINECS: 231-847-6
UN nummer: 3288
ADR: 6.1 III
Merck Index: 14,2653