Sølv(I)sulfid ≥99.9% (metals basis)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Disilver sulphide

89473.06 89473.14
89473.06EA 1040 DKK
89473.06 89473.14
Sølv(I)sulfid ≥99.9% (metals basis)
Sølv(I)sulfid

Purity: 99.9+% CAS No.: 21548-73-2 Molecular Formula: Ag2S Formula Weight: 247.80 Physical Form: Crystalline powder Density: 7.317 Appearance: Dark grey to black Odour: Odourless Melting Point: 825° Merck Reference: 14,8530 UN No.: 3077 MDL No.: MFCD00003406

Formel: Ag₂S
Molekylvægt: 247.8 g/mol
Smeltepunkt: 825 °C
Massefylde: 7.317
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003406
CAS-nummer: 21548-73-2
EINECS: 244-438-2
UN nummer: 3077
ADR: 9 III
Merck Index: 14,8530

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR