Vandopløselige papiretiketter B-403 til BBP30 eller 76 mm kerneprintere
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
149271 149268 149269 149270 149274 149275 149277 198760 198763 198762 198758 198759 198764
116-0788EA 1900 DKK
116-0788 116-0785 116-0786 116-0787 116-0789 116-0790 116-0791 116-0795 116-0796 116-0797 116-0793 116-0794 116-0792
Vandopløselige papiretiketter B-403 til BBP30 eller 76 mm kerneprintere
Etiketter
These water dissolvable paper labels are ideal to label items that need new identification after every use. Exposing the label to warm water will completely dissolve it within 30 seconds, without damage to the surface it was applied to, and without leaving any residue.
  • Self-adhesive
  • Extremely easy to apply
  • Adheres well to glass, stainless steel, aluminium, plastic and others
The labels reduce the labour required to remove labels, reduce maintenance to filters and drains and enable easy re-identification of reusable items in a range of industries.