ESD safety shoes, lace-up, jo_Soft Low, Jori
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
12751/47 12751/40 12751/41 12751/42 12751/43 12751/44 12751/45 12751/46 12751/48
ELTN12751/47EA 440 DKK
ELTN12751/47 ELTN12751/40 ELTN12751/41 ELTN12751/42 ELTN12751/43 ELTN12751/44 ELTN12751/45 ELTN12751/46 ELTN12751/48
ESD safety shoes, lace-up, jo_Soft Low, Jori
Sko og støvler
Leather-free safety shoes made from seamless hydrophobised CORDURA® (tear and abrasion resistant, dirt and water repellent), skin friendly textile lining, a full padded bellows tongue and collar padding plus a jo_TIGHT dual-density sole with profile (4,2 mm; oil, fuel and abrasion resistant, also temperature resistant from –20 to +130 °C).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Heel pocket lining made from sturdy abrasion resistant microfibre lining
  • Full length ESD inlay sole
  • PU scuff cap protects the upper material
  • Metal-free puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/red
Certificeringer: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.