Ytterbium(III)oxid ≥99.99% (REO, rare earth oxide basis), REacton®
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Diytterbium trioxide, Ytterbium oxide
11192.09
11192.18
11192.09EA
1320
DKK
11192.09
11192.18
Ytterbium(III)oxid ≥99.99% (REO, rare earth oxide basis), REacton®
Ytterbium(III) oxide
Purity: 99.99%
CAS No.: 1314-37-0
Molecular Formula: Yb2O3
Formula Weight: 394.08
Physical Form: Powder
Density: 9.2
Melting Point: 2227°
Merck Reference: 14,10105
MDL No.: MFCD00011290
Formel:
O₃Yb₂ Smeltepunkt: 2227 °C Massefylde: 9,2 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011290 CAS-nummer: 1314-37-0 EINECS: 215-234-0 Merck Index: 14,10105 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...