Safety shoes, lace-up, Basic Compo Low, Jori
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
12111/36 12111/37 12111/38 12111/39 12111/40 12111/41 12111/42 12111/43 12111/44 12111/45 12111/46 12111/47 12111/48
ELTN12111/36EA 321 DKK
ELTN12111/36 ELTN12111/37 ELTN12111/38 ELTN12111/39 ELTN12111/40 ELTN12111/41 ELTN12111/42 ELTN12111/43 ELTN12111/44 ELTN12111/45 ELTN12111/46 ELTN12111/47 ELTN12111/48
Safety shoes, lace-up, Basic Compo Low, Jori
Sko og støvler
100% metal-free, extremely lightweight safety shoes made from cowhide leather, reflective materials, skin friendly textile lining, a full padded bellows tongue and collar padding plus a dual-density PU/PU jo_Basic sole (oil, fuel and abrasion resistant, also heat resistant up to approx. 130 °C).
  • Climate-regulating, breathable textile lining
  • Full length inlay sole
  • PU scuff cap protects the upper material
  • Puncture resistant composite midsole
  • Protective composite toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects as well as areas close to induction loops or metal detectors.
Colour: Black
Certificeringer: EN ISO 20345; S3, SRC.