Powered air respirator (PAPR) system, Versaflo™ Turbo TR-300+
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TR3600 TR302EPLUS TR315EPLUS
111-2408EA 3130 DKK
111-2408 111-2404 111-2405
Powered air respirator (PAPR) system, Versaflo™ Turbo TR-300+
Åndedrætsværn Åndedrætsbeskyttelsessystemer
The balanced and compact 3M™ Versaflo™ TR-300+ is an easy-to-use, versatile respirator system for particulate environments.
  • Two selectable flow rate options: 185 and 205 L/min
  • Highly efficient filter captures hazardous dust, mist and fume plus options for nuisance level gases and vapours
  • Slim, lightweight balanced design for outstanding comfort
  • Audible and LED alarm signals low battery charge or low airflow
  • Lithium ion battery with LED to indicate charge status
  • Display shows battery charge status, selected airflow and particulate filter loading status during use
The TR-300+ Powered Air Turbo, when used as part of an approved system with appropriate filters, provides protection against:
Particles e.g. fine dusts, mists and metal fume.
Nuisance odour levels (below the Workplace Exposure Limit) of organic gas and vapours
Nuisance odour levels (below the Workplace Exposure Limit) of acid gases and up to 10× Workplace Exposure Limit of hydrogen fluoride.
Certificeringer: The TR-300+ Powered Air Turbo, in combination with 3M approved headtops, has been tested and approved to EN 12941:1998+A2:2008. Also meets the dust protection and spraying water requirements (IP 53) of EN 60529.
Leveringsinformation: TR-302E+ Turbo unit is supplied with filter cover, airflow indicator and spark arrestors.
TR-315E+ Starter Kit includes TR-302E+ turbo unit, particulate filter, standard belt, high capacity battery pack, battery charger kit, self-adjusting breathing tube, airflow indicator and spark arrestors.