ESD safety shoes, sandals, Velcro® fastening, Safety-Grip, White Easy Low ESD S1
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
72015/44 72015/40 72015/46 72015/45 72015/39 72015/43 72015/48 72015/42 72015/41 72015/47
114-8861EA 559 DKK
114-8861 114-8857 114-8863 114-8862 114-8856 114-8860 114-8865 114-8859 114-8858 114-8864
ESD safety shoes, sandals, Velcro® fastening, Safety-Grip, White Easy Low ESD S1
Sko og støvler
Leather-free, lightweight ESD safety sandal made from hydrophobised microfibre upper, a padded tongue and a slip resistant MONO-PU sole SAFETY-GRIP.
  • Climate regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full-length inlay sole ESD PRO grey
  • Anatomically shaped insole and ankle support for improved stability
  • Soft fleece ESD insole
  • Protective steel toecap (200 J)
Ideal for use in dry work areas such as industry, storage, transport, construction etc.
Colour: White
Certificeringer: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S1, SRC