Europium(III)oxid ≥99.996% (REO, rare earth oxide basis), REacton®

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

44347.06 44347.14 44347.22
44347.06EA 2700 DKK
44347.06 44347.14 44347.22
Europium(III)oxid ≥99.996% (REO, rare earth oxide basis), REacton®
Europium(III) oxide

Purity: 99.996% CAS No.: 1308-96-9 Molecular Formula: Eu2O3 Formula Weight: 351.92 Physical Form: Powder Sensitivity: Hygroscopic Merck Reference: 14,3904 MDL No.: MFCD00010997

Formel: Eu₂O₃
Molekylvægt: 351.92 g/mol
Smeltepunkt: 2291 °C
Massefylde: 7.42 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010997
CAS-nummer: 1308-96-9
EINECS: 215-165-6
Merck Index: 14,3904

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR