Europium(III)oxid ≥99.9% (REO, rare earth oxide basis), REacton®
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
11300.06
11300.14
11300.22
11300.06EA
1680
DKK
11300.06
11300.14
11300.22
Europium(III)oxid ≥99.9% (REO, rare earth oxide basis), REacton®
Europium(III) oxide
Purity: 99.9%
CAS No.: 1308-96-9
Molecular Formula: Eu2O3
Formula Weight: 351.92
Physical Form: Powder
Sensitivity: Hygroscopic
Merck Reference: 14,3904
MDL No.: MFCD00010997
Formel:
Eu₂O₃ Molekylvægt: 351.92 g/mol Smeltepunkt: 2291 °C Massefylde: 7.42 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00010997 CAS-nummer: 1308-96-9 EINECS: 215-165-6 Merck Index: 14,3904 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...