Dysprosium(III)oxid ≥99.9%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Didysprosium trioxide, Dysprosium oxide

36370.14 36370.22
36370.14EA 2470 DKK
36370.14 36370.22
Dysprosium(III)oxid ≥99.9%
Dysprosium(III) oxide

Purity: 99.9% CAS No.: 1308-87-8 Molecular Formula: Dy2O3 Formula Weight: 373.00 Physical Form: 3-12mm Sintered Lump Density: 7.81 Melting Point: 2340° Merck Reference: 14,3482 MDL No.: MFCD00010986

Formel: Dy₂O₃
Molekylvægt: 373 g/mol
Smeltepunkt: 2340 °C
Massefylde: 7.81
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010986
CAS-nummer: 1308-87-8
EINECS: 215-164-0
Merck Index: 14,3482

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR