Holmiumtrifluorid, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Holmium (III) fluoride
11278.06
11278.14
11278.06EA
1280
DKK
11278.06
11278.14
Holmiumtrifluorid, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®
Holmiumtrifluorid
Purity: 99.99%
CAS No.: 13760-78-6
Molecular Formula: HoF3
Formula Weight: 221.93
Physical Form: Crystalline Lump
Density: 7.6
Melting Point: 1143°
UN No.: 3288
MDL No.: MFCD00016142
Formel:
HoF₃ Molekylvægt: 221.93 g/mol Smeltepunkt: 1143 Massefylde: 7.6 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00016142 CAS-nummer: 13760-78-6 EINECS: 237-352-1 UN nummer: 3288 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...