Europium(III)fluorid, anhydrous ≥99.98% (REO, rare earth oxide basis), REacton®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
11302.04 11302.06 11302.14
11302.04EA 1630 DKK
11302.04 11302.06 11302.14
Europium(III)fluorid, anhydrous ≥99.98% (REO, rare earth oxide basis), REacton®
Europium(III) fluoride
Purity: 99.98% CAS No.: 13765-25-8 Molecular Formula: EuF3 Formula Weight: 208.96 Physical Form: Powder Sensitivity: Hygroscopic Boiling Point: 2280° MDL No.: MFCD00010998
Formel: EuF₃
Molekylvægt: 208.96 g/mol
Kogepunkt: >2280 °C
Smeltepunkt: 1390 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010998
CAS-nummer: 13765-25-8
EINECS: 237-368-9