Ceriumtrichlorid, ultra dry ≥99.9% (REO, rare earth oxide basis)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Cerous chloride, Cerium(III) chloride, Cerium (III) chloride

Ampuled under argon

35688.06 35688.14 35688.22
35688.06EA 1400 DKK
35688.06 35688.14 35688.22
Ceriumtrichlorid, ultra dry ≥99.9% (REO, rare earth oxide basis)
Ceriumtrichlorid

Purity: 99.9% CAS No.: 7790-86-5 Molecular Formula: CeCl3 Formula Weight: 246.48 Physical Form: Crystalline, Ampouled under argon Density: 3.92 Appearance: White to pale grey Odour: Odourless Melting Point: 848° Boiling Point: 1727° Merck Reference: 14,1997 MDL No.: MFCD00010929

Formel: CeCl₃
Molekylvægt: 246.47 g/mol
Kogepunkt: 1727 °C
Smeltepunkt: 848
Massefylde: 3.92
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010929
CAS-nummer: 7790-86-5
EINECS: 232-227-8
Merck Index: 14,1997

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR