Gadolinium tribromid, ultra dry ≥99.99% (metals basis)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel


Ampuled under argon

43673.14 43673.06
43673.14EA 7120 DKK
43673.14 43673.06
Gadolinium tribromid, ultra dry ≥99.99% (metals basis)
Gadolinium tribromid

Purity: 99.99% CAS No.: 13818-75-2 Molecular Formula: GdBr3 Formula Weight: 396.96 Physical Form: Powder, Ampouled under argon Sensitivity: Hygroscopic MDL No.: MFCD00049741

Formel: Br₃Gd
Smeltepunkt: 770 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00049741
CAS-nummer: 13818-75-2
EINECS: 237-494-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR