2-Phenylindol 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
130680050
130681000
ACRO130680050EA
239
DKK
ACRO130680050
ACRO130681000
2-Phenylindol 99%
2-Phenylindole
Formel:
C₁₄H₁₁N Kogepunkt: 250 °C (10 mmHg) Smeltepunkt: 188…191 °C |
MDL Number:
MFCD00005608 CAS-nummer: 948-65-2 EINECS: 213-436-3 |
Specifikationer Testresultater
Appearance | Off-white to beige to light green Powder |
Infrared spectrum | Conforms |
Melting point | 188 to 191 °C |
Water | ≤0.5 % (K.F.) |
Sulfated ash | ≤0.2 % |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...