5-Methylindol ≥99%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Purity: 99%
CAS No.: 614-96-0
Molecular Formula: C9H9N
Formula Weight: 131.18
Storage Temperature: 2°C to 8°C
Melting Point: 58-61°
Boiling Point: 100°/3mm
Beilstein Reference: 2228
MDL No.: MFCD00005680
Formel:
C₉H₉N Kogepunkt: 100 °C (3 mmHg) Smeltepunkt: 58…61 °C Massefylde: 1,0202 g/cm³ (78.3 °C) |
MDL Number:
MFCD00005680 CAS-nummer: 614-96-0 EINECS: 210-400-9 |