2-Methylindol 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Methyl-1H-indole
127021000
127025000
ACRO127021000EA
577
DKK
ACRO127021000
ACRO127025000
2-Methylindol 98%
2-Methylindole
Formel:
C₉H₉N Molekylvægt: 131.18 g/mol Kogepunkt: 273 °C (760 mmHg) Smeltepunkt: 56…60 °C Flammepunkt: 141 °C Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00005616 CAS-nummer: 95-20-5 EINECS: 202-398-3 UN nummer: 3335 ADR: 9 III |
Specifikationer Testresultater
Appearance | Yellow to red-purple to brown Crystalline powder or crystals or flakes |
Infrared spectrum | Conforms |
Melting point | 56 to 60 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...