6-Fluorindol 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

342510010 342512500 342510050
ACRO342510010EA 406 DKK
ACRO342510010 ACRO342512500 ACRO342510050
6-Fluorindol 98%
6-Fluoroindole
Formel: C₈H₆FN
Smeltepunkt: 72…77 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00056933
CAS-nummer: 399-51-9

Bestil nu

Specifikationer Testresultater

Appearance Beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 73 to 77 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR