Indol-3-carboxylsyre 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1H-Indole-3-carboxylic acid
307610010
307610050
ACRO307610010EA
313
DKK
ACRO307610010
ACRO307610050
Indol-3-carboxylsyre 99%
Indol-3-carboxylsyre
Formel:
C₉H₇NO₂ Smeltepunkt: 200…203 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00005624 CAS-nummer: 771-50-6 EINECS: 212-231-6 |
Specifikationer Testresultater
Appearance | Light beige Powder |
Infrared spectrum | Conforms |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...