Inden ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1H-Indene

L12665.06 L12665.14 L12665.22
L12665.06EA 880 DKK
L12665.06 L12665.14 L12665.22
Inden ≥97%
Inden

Purity: 97% CAS No.: 95-13-6 Molecular Formula: C9H8 Formula Weight: 116.16 Storage Temperature: 2°C to 8°C Flash Point: 58°(136°F) Refractive Index: 1.5740 Odour: Aromatic Melting Point: ca -2° Boiling Point: 181-182° Beilstein Reference: 635873 Merck Reference: 14,4939 UN No.: 3295 MDL No.: MFCD00003777

Formel: C₉H₈
Molekylvægt: 116.16 g/mol
Kogepunkt: 181…182 °C
Smeltepunkt: ∼–2 °C
Massefylde: 0.996
Flammepunkt: 58 °C (136 °F)
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00003777
CAS-nummer: 95-13-6
EINECS: 202-393-6
UN nummer: 3295
ADR: 3 III
Merck Index: 13,04964

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR