Ninhydrin
Leverandør: Serva
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
30410.01
30410.02
SERA30410.01EA
0
DKK
SERA30410.01
SERA30410.02
Ninhydrin
Ninhydrin
Formel:
C₉H₆O₄ Molekylvægt: 178.14 g/mol Kogepunkt: 351 Smeltepunkt: 250…258 °C Massefylde: 0.862 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003791 CAS-nummer: 485-47-2 EINECS: 207-618-1 Merck Index: 13,06582 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...