5-Brom-1-indanon ≥97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
5-Bromo-2,3-dihydro-1H-inden-1-one
B25707.03
B25707.06
B25707.03EA
473
DKK
B25707.03
B25707.06
5-Brom-1-indanon ≥97%
5-Brom-1-indanon
Purity: 98%
CAS No.: 34598-49-7
Molecular Formula: C9H7BrO
Formula Weight: 211.06
Melting Point: 125-129°
Beilstein Reference: 2357199
Formel:
C₉H₇BrO Kogepunkt: 280 °C (1013 hPa) Smeltepunkt: 125…129 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00082718 CAS-nummer: 34598-49-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...