2-Ethylimidazol ≥98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 98%
CAS No.: 1072-62-4
Molecular Formula: C5H8N2
Formula Weight: 96.13
Flash Point: 159°(318°F)
Melting Point: 78-86°
Boiling Point: 268°
Beilstein Reference: 106381
MDL No.: MFCD00005192
Formel:
C₅H₈N₂ Kogepunkt: 268 °C Smeltepunkt: 78…86 °C Massefylde: 1,03 g/cm³ (25 °C) Flammepunkt: 159 °C (318 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00005192 CAS-nummer: 1072-62-4 EINECS: 214-011-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...