Griess reagent R2
Leverandør: Cayman Chemical
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
780020-8 780020-50
CAYM780020-8EA 426 DKK
CAYM780020-8 CAYM780020-50
Griess reagent R2
Griess reagent
Griess Reagent component for determination of nitrite in aqueous solutions.