Dimethylsulfoxid, dry_extra 99.7+%, AcroSeal®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
326881000 326880010 326880025
326881000.EA 704 DKK
326881000. 326880010. 326880025.
Dimethylsulfoxid, dry_extra 99.7+%, AcroSeal®
Dimethylsulfoxid
Formel: (CH₃)₂SO
Molekylvægt: 78.14 g/mol
Kogepunkt: 189 °C (760 mmHg)
Smeltepunkt: 18.4 °C
Massefylde: 1.1 g/cm³
Flammepunkt: 87 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002089
CAS-nummer: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.70 %
Residue after evaporation ≤0.001 %