Sulfamidsyre, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel


M-Clarity™ quality level = MQ200

1.00219.2500 1.00219.9025
1.00219.2500EA 1100 DKK
1.00219.2500 1.00219.9025
Sulfamidsyre, Supelco®
Sulfamidsyre
Formel: H₃NO₃S
Kogepunkt: 247 °C (1013 hPa)
Smeltepunkt: 205 °C
Massefylde: 2,15 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011603
CAS-nummer: 5329-14-6
EINECS: 226-218-8
UN nummer: 2967
ADR: 8,III
Merck Index: 13,09006
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afa0500786be69d531cdf06f8e67548e6f67b23f&vid=73aa20196fbabf965e4375f2cf3675e2f513ee59&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Solubility 213 g/l (20 °C)
Melting Point 205 °C (decomposition)
Molar Mass 97.08 g/mol
Bulk Density 600 kg/m3
Density 2.13 g/cm3 (20 °C)
pH 1.2 (10 g/l, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR