Sulfamidsyre ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Sulfaminsyre
A18836.0E
A18836.22
A18836.36
A18836.0EEA
1320
DKK
A18836.0E
A18836.22
A18836.36
Sulfamidsyre ≥98%
Sulfamidsyre
Purity: 98+%
CAS No.: 5329-14-6
Molecular Formula: H3NO3S
Formula Weight: 97.09
Odour: Odourless
pH: 1.2
Melting Point: ca 205° dec.
Merck Reference: 14,8921
UN No.: 2967
MDL No.: MFCD00011603
Formel:
H₃NO₃S Molekylvægt: 97.09 g/mol Kogepunkt: 247 Smeltepunkt: ∼ 205 °C (dec.) Massefylde: 2.12 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011603 CAS-nummer: 5329-14-6 EINECS: 226-218-8 UN nummer: 2967 ADR: 8 III Merck Index: 13,09006 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...