Triphenylarsin ≥97%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 97%
CAS No.: 603-32-7
Molecular Formula: C18H15As
Formula Weight: 306.24
Flash Point: 265°(509°F)
Melting Point: 58-62°
Boiling Point: 360°
Beilstein Reference: 1842037
UN No.: 3465
MDL No.: MFCD00002994
Formel:
C₁₈H₁₅As Kogepunkt: 360 °C Smeltepunkt: 58…62 °C Massefylde: 1,306 g/cm³ (20 °C) Flammepunkt: 265 °C (509 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002994 CAS-nummer: 603-32-7 EINECS: 210-032-9 UN nummer: 3465 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...