Tetrabutylammoniumnitrat 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
420130250
420130050
420131000
ACRO420130250EA
955
DKK
ACRO420130250
ACRO420130050
ACRO420131000
Tetrabutylammoniumnitrat 98%
Tetrabutylammonium nitrate
Formel:
C₁₆H₃₆N₂O₃ Smeltepunkt: 116…120 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00043202 CAS-nummer: 1941-27-1 EINECS: 217-726-0 UN nummer: 1479 ADR: 5.1,II |
Specifikationer Testresultater
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥97.5 % |
HPLC | ≥88 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...