Tetramethylammoniumiodid ≥99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
A12811.22
A12811.36
A12811.22EA
1450
DKK
A12811.22
A12811.36
Tetramethylammoniumiodid ≥99%
Tetramethylammonium iodide
Purity: 99%
CAS No.: 75-58-1
Molecular Formula: C4H12IN
Formula Weight: 201.05
Odour: Odourless
Melting Point:
Formel:
C₄H₁₂IN Smeltepunkt: >300 °C Massefylde: 1,84 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011629 CAS-nummer: 75-58-1 EINECS: 200-881-3 UN nummer: 2811 ADR: 6.1,III Merck Index: 14,9225 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...