Tetrabutylammoniumfluorid trihydrat 99%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
221080100 221080500 221082500
ACRO221080100EA 381 DKK
ACRO221080100 ACRO221080500 ACRO221082500
Tetrabutylammoniumfluorid trihydrat 99%
Tetrabutylammoniumfluorid trihydrat
Formel: C₁₆H₃₆FN·3H₂O
Molekylvægt: 315.51 g/mol
Smeltepunkt: 58…60 °C
MDL Number: MFCD00149981
CAS-nummer: 87749-50-6
EINECS: 207-057-2
UN nummer: 1759
ADR: 8 III
Bestil nu
Specifikationer Testresultater
Appearance White to light yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %