Tetrabutylammonium chlorid, Supelco®
Leverandør: Merck
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
M-Clarity™ quality level = MQ100
86852-10G-F
86852-50G-F
86852-10G-FEA
2630
DKK
86852-10G-F
86852-50G-F
Tetrabutylammonium chlorid, Supelco®
Tetrabutylammonium chlorid
Formel:
C₁₆H₃₆ClN Molekylvægt: 277.92 g/mol Smeltepunkt: 83…86 °C Massefylde: 1.05 g/cm³ (20 °C) Flammepunkt: 110 |
MDL Number:
MFCD00011635 CAS-nummer: 1112-67-0 EINECS: 214-195-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...