Tetraheptylammoniumbromid, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
M-Clarity™ quality level = MQ100
87296-10G-F 87296-50G-F
87296-10G-FEA 1150 DKK
87296-10G-F 87296-50G-F
Tetraheptylammoniumbromid, Supelco®
Tetraheptylammonium bromide
Formel: C₂₈H₆₀BrN
Molekylvægt: 490.7 g/mol
Smeltepunkt: 96
MDL Number: MFCD00011861
CAS-nummer: 4368-51-8
EINECS: 224-459-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b02cb266f9a1f8dd8d5d5e4f11ad8c69e47b90b&vid=7691c2e7f3a8e02e879853712adec7b46378801d&prid=web&clid=SAPDAM