Diarsenictriselenid ≥99.999% (metals basis)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Arsenic(III) selenide
43748.14
43748.06
43748.14EA
9560
DKK
43748.14
43748.06
Diarsenictriselenid ≥99.999% (metals basis)
Diarsenictriselenid
Purity: 99.999%
CAS No.: 1303-36-2
Molecular Formula: As2Se3
Formula Weight: 386.72
Physical Form: -160 Mesh Powder
Density: 4.75
Melting Point: 260°
Merck Reference: 14,805
UN No.: 3077
MDL No.: MFCD00014166
Formel:
As₂Se₃ Smeltepunkt: 260 °C Massefylde: 4,75 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00014166 CAS-nummer: 1303-36-2 EINECS: 215-119-5 UN nummer: 3077 ADR: 9,III Merck Index: 14,805 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...