Bismut (III) oxid ≥99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Bismuth Trioxide, Dibismuth trioxide

12230.30 12230.A1 12230.A7
12230.30EA 820 DKK
12230.30 12230.A1 12230.A7
Bismut (III) oxid ≥99%
Bismut (III) oxid

Purity: 99% CAS No.: 1304-76-3 Molecular Formula: Bi2O3 Formula Weight: 465.96 Physical Form: Powder Density: 8.9 Appearance: Yellow Melting Point: 817° Boiling Point: ca 1890° Solubility: Practically insoluble in water. Soluble in acids Merck Reference: 14,1273 MDL No.: MFCD00003462


In disinfectants, magnets, glass, rubber, vulcanization, in fireproofing papers and polymers, and in catalysts

Formel: BiO₃
Molekylvægt: 465.96 g/mol
Kogepunkt: ∼1890 °C
Smeltepunkt: 817 °C
Massefylde: 8.9
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003462
CAS-nummer: 1304-76-3
EINECS: 215-134-7
Merck Index: 13,01273

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR