Safety spectacles, SecureFit™ 600
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SF602SGAF-EU SF650AS-EU SF601SGAF-EU SF603SGAF-EU SF617AS-EU SF630AS-EU SF601RAS-EU SF610AS-EU SF611AS-EU
111-2283EA 209 DKK
111-2283 111-2287 111-2282 111-2284 111-2285 111-2286 111-2289 111-2290 MMMASF611AS-EU
Safety spectacles, SecureFit™ 600
Briller
The 3M™ SecureFit™ SF600 series is an extension to the successful SecureFit range of protective eyewear. The innovative design promotes improved comfort, coverage and a good field of vision based on two lenses in a semi-rimless design.
  • Integrated side-shield
  • Temples feature 3M™ Pressure Diffusion technology which helps diffuse pressure over the ear to enhance frame comfort across a diverse workforce
  • Soft nose pads provide increased comfort and improved grip
  • Removable foam lined insert helps keep debris out of the eyes
  • Small vents in the insert can help reduce fogging
  • Optimal optical performance by having a dual lens design and optical class 1 lens
These products may be suitable for use in a wide range of applications including construction, engineering, general assembly, inspection work, light duty maintenance and repair, certain welding applications (1,7, 3,0 and 5,0 lens options), oil and gas industries, airports, workers at sea (polarized lens).
Weight: 27 g
Certificeringer: EN 166 FT CE; EN 170:2002 (clear and amber lenses), EN 172:1994 (5-1.7 and 5-3.1); EN 169:2002 (welding 1.7, 3.0, 5.0)