Tetrabutylgermanium ≥99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Tetra-n-butylgermane, Tetra-n-butylgermanium

34119.06 34119.14 34119.03
34119.06EA 1460 DKK
34119.06 34119.14 34119.03
Tetrabutylgermanium ≥99%
Tetrabutylgermanium

Purity: 99% CAS No.: 1067-42-1 Molecular Formula: [CH3(CH2)3]4Ge Formula Weight: 301.05 Flash Point: 110°(230°F) Physical Form: Liquid Density: 0.934 Refractive Index: 1.4571 Boiling Point: 277-279° MDL No.: MFCD00015224

Formel: C₁₆H₃₆Ge
Molekylvægt: 301.07 g/mol
Kogepunkt: 277…279 °C
Massefylde: 0.934
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00015224
CAS-nummer: 1067-42-1
EINECS: 213-929-3

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR